missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Aminocyclohexanecarboxylic acid, 98%
CAS: 2756-85-6 | C7H13NO2 | 143.19 g/mol
$64.08 - $206.53
Chemical Identifiers
| CAS | 2756-85-6 |
|---|---|
| Molecular Formula | C7H13NO2 |
| Molecular Weight (g/mol) | 143.19 |
| MDL Number | MFCD00001487 |
| InChI Key | WOXWUZCRWJWTRT-UHFFFAOYSA-N |
| Synonym | 1-aminocyclohexanecarboxylic acid, homocycloleucine, 1-amino-1-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-amino, alpha-aminocyclohexanecarboxylic acid, alpha-aminocyclohexanecarboxylic aicd, unii-1se3hd391h, 1-amino-1-cyclohexane carboxylic acid, .alpha.-aminocyclohexanecarboxylic acid, .alpha.-aminocyclohexanecarboxylic aicd |
| PubChem CID | 1366 |
| ChEBI | CHEBI:86534 |
| IUPAC Name | 1-aminocyclohexane-1-carboxylic acid |
| SMILES | NC1(CCCCC1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1185306
|
Thermo Scientific Chemicals
A1185306 |
5 g |
Each for $64.08
|
|
|||||
|
AAA1185314
|
Thermo Scientific Chemicals
A1185314 |
25 g |
Each for $206.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2756-85-6 | |
| 143.19 | |
| WOXWUZCRWJWTRT-UHFFFAOYSA-N | |
| 1366 | |
| 1-aminocyclohexane-1-carboxylic acid |
| C7H13NO2 | |
| MFCD00001487 | |
| 1-aminocyclohexanecarboxylic acid, homocycloleucine, 1-amino-1-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-amino, alpha-aminocyclohexanecarboxylic acid, alpha-aminocyclohexanecarboxylic aicd, unii-1se3hd391h, 1-amino-1-cyclohexane carboxylic acid, .alpha.-aminocyclohexanecarboxylic acid, .alpha.-aminocyclohexanecarboxylic aicd | |
| CHEBI:86534 | |
| NC1(CCCCC1)C(O)=O |
Specifications
| 2756-85-6 | |
| C7H13NO2 | |
| 5 g | |
| Hygroscopic | |
| WOXWUZCRWJWTRT-UHFFFAOYSA-N | |
| 1-aminocyclohexane-1-carboxylic acid | |
| 1366 | |
| 143.19 | |
| 1-Aminocyclohexanecarboxylic acid |
| >300°C | |
| MFCD00001487 | |
| 2355692 | |
| 1-aminocyclohexanecarboxylic acid, homocycloleucine, 1-amino-1-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-amino, alpha-aminocyclohexanecarboxylic acid, alpha-aminocyclohexanecarboxylic aicd, unii-1se3hd391h, 1-amino-1-cyclohexane carboxylic acid, .alpha.-aminocyclohexanecarboxylic acid, .alpha.-aminocyclohexanecarboxylic aicd | |
| NC1(CCCCC1)C(O)=O | |
| 143.19 | |
| CHEBI:86534 | |
| 98% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-411-0
RTECSNumber : GU8393000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only