missing translation for 'onlineSavingsMsg'
Learn More

1-Aminocyclohexanecarboxylic acid, 98%

Catalog No. AAA1185314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1185314 25 g
AAA1185306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1185314 Supplier Thermo Scientific Chemicals Supplier No. A1185314
Only null left

CAS: 2756-85-6 | C7H13NO2 | 143.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2756-85-6
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00001487
InChI Key WOXWUZCRWJWTRT-UHFFFAOYSA-N
Synonym 1-aminocyclohexanecarboxylic acid, homocycloleucine, 1-amino-1-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-amino, alpha-aminocyclohexanecarboxylic acid, alpha-aminocyclohexanecarboxylic aicd, unii-1se3hd391h, 1-amino-1-cyclohexane carboxylic acid, .alpha.-aminocyclohexanecarboxylic acid, .alpha.-aminocyclohexanecarboxylic aicd
PubChem CID 1366
ChEBI CHEBI:86534
IUPAC Name 1-aminocyclohexane-1-carboxylic acid
SMILES NC1(CCCCC1)C(O)=O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 2355692
Sensitivity Hygroscopic
Formula Weight 143.19
Percent Purity 98%
Chemical Name or Material 1-Aminocyclohexanecarboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-411-0
RTECSNumber GU8393000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only