missing translation for 'onlineSavingsMsg'
Learn More

1-Aminobenzotriazole 98.0+%, TCI America™

Catalog No. p-7117454
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14641G 1 g
A14645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14641G Supplier TCI America Supplier No. A14641G
Only null left

Chemical Identifiers

CAS 1614-12-6
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.142
MDL Number MFCD00132902
InChI Key JCXKHYLLVKZPKE-UHFFFAOYSA-N
Synonym 1-aminobenzotriazole, 1h-benzotriazol-1-amine, 1h-benzo d 1,2,3 triazol-1-amine, 1-benzotriazolamine, 1-abtz, 1h-1,2,3-benzotriazol-1-amine, 1h-1,2,3-benzotriazol-1-ylamine, 1,2,3-benzotriazol-1-amine, benzotriazolylamine, 1-benzotriazolylamine
PubChem CID 1367
IUPAC Name benzotriazol-1-amine
SMILES C1=CC=C2C(=C1)N=NN2N

Specifications

Melting Point 84°C
Quantity 1 g
Formula Weight 134.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminobenzotriazole
RTECSNumber DM1235000
TSCA No