missing translation for 'onlineSavingsMsg'
Learn More

1-Aminobenzotriazole, 98%

CAS: 1614-12-6 | C6H6N4 | 134.14 g/mol

$465.33 - $465.33

Chemical Identifiers

CAS 1614-12-6
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.14
MDL Number MFCD00132902
InChI Key JCXKHYLLVKZPKE-UHFFFAOYSA-N
Synonym 1-aminobenzotriazole, 1h-benzotriazol-1-amine, 1h-benzo d 1,2,3 triazol-1-amine, 1-benzotriazolamine, 1-abtz, 1h-1,2,3-benzotriazol-1-amine, 1h-1,2,3-benzotriazol-1-ylamine, 1,2,3-benzotriazol-1-amine, benzotriazolylamine, 1-benzotriazolylamine
PubChem CID 1367
IUPAC Name benzotriazol-1-amine
SMILES C1=CC=C2C(=C1)N=NN2N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC348642500
SDS View Documents
Thermo Scientific Chemicals
348642500
250 mg Glass bottle
Each for $465.33
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1614-12-6
134.14
JCXKHYLLVKZPKE-UHFFFAOYSA-N
1367
C1=CC=C2C(=C1)N=NN2N
C6H6N4
MFCD00132902
1-aminobenzotriazole, 1h-benzotriazol-1-amine, 1h-benzo d 1,2,3 triazol-1-amine, 1-benzotriazolamine, 1-abtz, 1h-1,2,3-benzotriazol-1-amine, 1h-1,2,3-benzotriazol-1-ylamine, 1,2,3-benzotriazol-1-amine, benzotriazolylamine, 1-benzotriazolylamine
benzotriazol-1-amine

Specifications

1614-12-6
100.0
Beige to Yellow or Brown
97.5% min. (HPLC)
C6H6N4
250 mg
Solubility in water: slightly soluble. Other solubilities: soluble in chloroform,carbon tetrachloride,,soluble in actic acid
C1=CC=C2C(=C1)N=NN2N
134.14
134.14
Crystalline Powder
97.5
81°C to 85°C
Authentic
Glass bottle
MFCD00132902
1-aminobenzotriazole, 1h-benzotriazol-1-amine, 1h-benzo d 1,2,3 triazol-1-amine, 1-benzotriazolamine, 1-abtz, 1h-1,2,3-benzotriazol-1-amine, 1h-1,2,3-benzotriazol-1-ylamine, 1,2,3-benzotriazol-1-amine, benzotriazolylamine, 1-benzotriazolylamine
JCXKHYLLVKZPKE-UHFFFAOYSA-N
benzotriazol-1-amine
1367
98%
1-Aminobenzotriazole, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only