missing translation for 'onlineSavingsMsg'
Learn More

1-Aminoanthraquinone 98.0+%, TCI America™

Catalog No. p-7116964
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A059025G 25 g
A0590500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A059025G Supplier TCI America Supplier No. A059025G
Only null left

Chemical Identifiers

CAS 82-45-1
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.231
MDL Number MFCD00001213
InChI Key KHUFHLFHOQVFGB-UHFFFAOYSA-N
Synonym 1-aminoanthraquinone, diazo fast red al, anthraquinone, 1-amino, 1-amino anthraquinone, 1-aminoanthrachinon, 1-amino-anthraquinone, 9,10-anthracenedione, 1-amino, alpha-aminoanthraquinone, 1-amino-9,10-anthraquinone, 1-amino-9,10-anthracenedione
PubChem CID 6710
IUPAC Name 1-aminoanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)N

Specifications

Melting Point 257°C
Quantity 25 g
UN Number 3077
Formula Weight 223.23
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminoanthraquinone
EINECSNumber (4)-0706
RTECSNumber CB5075000
TSCA Yes