missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-methylpiperazine 98.0+%, TCI America™

Catalog No. A119225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A119225ML 25 mL
A1192500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A119225ML Supplier TCI America Supplier No. A119225ML

Chemical Identifiers

CAS 6928-85-4
Molecular Formula C5H13N3
Molecular Weight (g/mol) 115.18
MDL Number MFCD00006154
InChI Key RJWLLQWLBMJCFD-UHFFFAOYSA-N
Synonym 1-amino-4-methylpiperazine, 1-piperazinamine, 4-methyl, 4-methyl-1-piperazinamine, 4-methyl-piperazin-1-ylamine, 1-methyl-4-aminopiperazine, 4-amino-1-methylpiperazine, 4-methylpiperazinylamine, 1-methyl-4-aminopierazine, pubchem8577, n-amino-4-methylpiperazine
PubChem CID 81349
IUPAC Name 4-methylpiperazin-1-amine
SMILES CN1CCN(CC1)N

Specifications

Color Yellow
Boiling Point 175°C
Quantity 25 mL
Formula Weight 115.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Amino-4-methylpiperazine