missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-methylpiperazine, 98%

Catalog No. AAL1910814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1910814 25 g
AAL1910822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1910814 Supplier Thermo Scientific Chemicals Supplier No. L1910814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6928-85-4 | C5H13N3 | 115.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6928-85-4
Molecular Formula C5H13N3
Molecular Weight (g/mol) 115.18
MDL Number MFCD00006154
InChI Key RJWLLQWLBMJCFD-UHFFFAOYSA-N
Synonym 1-amino-4-methylpiperazine, 1-piperazinamine, 4-methyl, 4-methyl-1-piperazinamine, 4-methyl-piperazin-1-ylamine, 1-methyl-4-aminopiperazine, 4-amino-1-methylpiperazine, 4-methylpiperazinylamine, 1-methyl-4-aminopierazine, pubchem8577, n-amino-4-methylpiperazine
PubChem CID 81349
IUPAC Name 4-methylpiperazin-1-amine
SMILES CN1CCN(CC1)N

Specifications

Density 0.96
Boiling Point 172°C to 175°C
Flash Point 70°C (158°F)
Refractive Index 1.484
Quantity 25 g
UN Number UN2735
Beilstein 103334
Sensitivity Air and light sensitive
Formula Weight 115.18
Percent Purity 98%
Chemical Name or Material 1-Amino-4-methylpiperazine
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 230-053-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only