missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-hydroxyanthraquinone, 96%

Catalog No. AAB2438806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2438806 5 g
AAB2438814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2438806 Supplier Thermo Scientific Chemicals Supplier No. B2438806

CAS: 116-85-8 | C14H9NO3 | 239.23 g/mol

The use of the inclusion complex of 1-amino-4-hydroxyanthraquinone (AHA) in the internal cavity of β-cyclodextrin confers increased sensitivity, selectivity and detection limit, and allows the determination of 10-70 ng ml-1 beryllium, compared to 60-500 ng ml-1 in the absence of the cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The use of the inclusion complex of 1-amino-4-hydroxyanthraquinone (AHA) in the internal cavity of β-cyclodextrin confers increased sensitivity, selectivity and detection limit, and allows the determination of 10-70 ng ml-1 beryllium, compared to 60-500 ng ml-1 in the absence of the cyclodextrin.

Solubility
Soluble in water, ethanol, acetone, benzene.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 116-85-8
Molecular Formula C14H9NO3
Molecular Weight (g/mol) 239.23
MDL Number MFCD00001223
InChI Key AQXYVFBSOOBBQV-UHFFFAOYSA-N
Synonym 1-amino-4-hydroxyanthraquinone, disperse red 15, calcosyn pink b, celutate pink b, nacelan pink b, amacel pink b, celutate pink bn, celutate pink by, cibacete red 3b, disperse red 2s
PubChem CID 8323
IUPAC Name 1-amino-4-hydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)N

Specifications

Melting Point 207°C to 209°C
Quantity 5 g
Solubility Information Soluble in water,ethanol,acetone,benzene.
Formula Weight 239.23
Percent Purity 96%
Chemical Name or Material 1-Amino-4-hydroxyanthraquinone
Hazard Category H304-H341-H351
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P281-P301+P310-P308+P313-P331-P501c
EINECSNumber 204-163-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only