missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-cyclopentylpiperazine, 97%

Catalog No. p-7036524
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54831MD 250 mg
AAH5483103 1 g
AAH5483106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54831MD Supplier Thermo Scientific Chemicals Supplier No. H54831MD

CAS: 61379-64-4 | C9H19N3 | 169.272 g/mol

1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 61379-64-4
Molecular Formula C9H19N3
Molecular Weight (g/mol) 169.272
MDL Number MFCD00190598
InChI Key QYHRIASMJNLWHJ-UHFFFAOYSA-N
PubChem CID 1512485
IUPAC Name 4-cyclopentylpiperazin-1-amine
SMILES C1CCC(C1)N2CCN(CC2)N

Specifications

Quantity 250 mg
Synonym 1-amino-4-cyclopentylpiperazine, 1-amino-4-cyclopentyl piperazine, 4-cyclopentyl-1-piperazinamine, pubchem8581, 4-cyclopentylpiperazinylamine, 1-piperazinamine,4-cyclopentyl, 1-piperazinamine, 4-cyclopentyl, 4-cyclopentyl-piperazin-1-ylamine, 1-n-cyclopentyl piperazine-4-amine, 1-piperazinamine, 4-cyclopentyl-9ci
Solubility Information Slightly soluble in water.
Formula Weight 169.27
Percent Purity 97%
Chemical Name or Material 1-Amino-4-cyclopentylpiperazine
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
EINECSNumber 262-742-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only