missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Amino-4-bromonaphthalene 98.0+%, TCI America™
$85.75 - $296.38
Chemical Identifiers
| CAS | 2298-07-9 |
|---|---|
| Molecular Formula | C10H8BrN |
| Molecular Weight (g/mol) | 222.09 |
| InChI Key | LIUKLAQDPKYBCP-UHFFFAOYSA-N |
| Synonym | 4-Bromo-1-naphthylamine |
| PubChem CID | 75303 |
| IUPAC Name | 4-bromonaphthalen-1-amine |
| SMILES | C1=CC=C2C(=C1)C(=CC=C2Br)N |
Chemical Identifiers
| 2298-07-9 | |
| 222.09 | |
| 4-Bromo-1-naphthylamine | |
| 4-bromonaphthalen-1-amine |
| C10H8BrN | |
| LIUKLAQDPKYBCP-UHFFFAOYSA-N | |
| 75303 | |
| C1=CC=C2C(=C1)C(=CC=C2Br)N |
Specifications
| 2298-07-9 | |
| Purple-White | |
| 5 g | |
| LIUKLAQDPKYBCP-UHFFFAOYSA-N | |
| 4-bromonaphthalen-1-amine | |
| 75303 | |
| ≥98.0% (GC) | |
| 1-Amino-4-bromonaphthalene |
| 102°C | |
| C10H8BrN | |
| 4-Bromo-1-naphthylamine | |
| C1=CC=C2C(=C1)C(=CC=C2Br)N | |
| 222.09 | |
| 222.09 | |
| Crystalline Powder |
Safety and Handling
TSCA : No