missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-bromoanthraquinone-2-sulfonic Acid Sodium Salt 98.0+%, TCI America™

Catalog No. A027925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A027925G 25 g
A0279500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A027925G Supplier TCI America Supplier No. A027925G

Chemical Identifiers

CAS 6258-06-6
Molecular Formula C14H7BrNNaO5S
Molecular Weight (g/mol) 404.17
MDL Number MFCD00019160
InChI Key TXMRAEGWZZVGIH-UHFFFAOYSA-M
Synonym Bromaminic Acid Sodium Salt
PubChem CID 5076555
IUPAC Name sodium;1-amino-4-bromo-9,10-dioxoanthracene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C=C(C(=C3C2=O)N)S(=O)(=O)[O-])Br.[Na+]

Specifications

Quantity 25 g
Formula Weight 404.17
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-4-bromoanthraquinone-2-sulfonic Acid Sodium Salt
EINECSNumber (4)-0701
RTECSNumber CB1090000
TSCA Yes