missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic Acid 95.0+%, TCI America™

Catalog No. A2526100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2526100MG 100 mg
A2526500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2526100MG Supplier TCI America Supplier No. A2526100MG
Only null left

Chemical Identifiers

CAS 905954-28-1
Molecular Formula C15H31NO8
Molecular Weight (g/mol) 353.412
InChI Key PVRGRRPCHFTMMD-UHFFFAOYSA-N
Synonym 21-Amino-4,7,10,13,16,19-hexaoxahenicosanoic Acid, Amino-PEG6-carboxylic Acid, Carboxy-PEG6-amine
PubChem CID 51035062
IUPAC Name 3-[2-[2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]propanoic acid
SMILES C(COCCOCCOCCOCCOCCOCCN)C(=O)O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 353.41
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic Acid