missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-3,3-diethoxypropane, 97%

CAS: 41365-75-7 | C7H17NO2 | 147.22 g/mol

$318.46 - $318.46

Chemical Identifiers

CAS 41365-75-7
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.22
InChI Key PXXMSHBZYAOHBD-UHFFFAOYSA-N
Synonym 1-amino-3,3-diethoxypropane, 3,3-diethoxypropylamine, 3-aminopropionaldehyde diethylacetal, 1-propanamine, 3,3-diethoxy, 3-aminopropionaldehyde diethyl acetal, propionaldehyde, 3-amino-, diethyl acetal, zlchem 1268, acmc-1apws, 3,3-diethoxy-propylamine, 3,3-diethoxypropyl amine
PubChem CID 521029
IUPAC Name 3,3-diethoxypropan-1-amine
SMILES CCOC(CCN)OCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC269850250
SDS View Documents
Thermo Scientific Chemicals
269850250
25 g Glass bottle
Each for $318.46
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

41365-75-7
147.22
1-amino-3,3-diethoxypropane, 3,3-diethoxypropylamine, 3-aminopropionaldehyde diethylacetal, 1-propanamine, 3,3-diethoxy, 3-aminopropionaldehyde diethyl acetal, propionaldehyde, 3-amino-, diethyl acetal, zlchem 1268, acmc-1apws, 3,3-diethoxy-propylamine, 3,3-diethoxypropyl amine
3,3-diethoxypropan-1-amine
C7H17NO2
PXXMSHBZYAOHBD-UHFFFAOYSA-N
521029
CCOC(CCN)OCC

Specifications

41365-75-7
0.9100g/mL
78°C
97%
C7H17NO2
(C2H5O)2CHCH2CH2NH2
0.91
Solubility in water: miscible, hydrolysis. Other solubilities: miscible with alcohol and petroleum ether
CCOC(CCN)OCC
147.22
147.22
Liquid
Colorless to Yellow
72°C (16.0 hPa)
Authentic
Glass bottle
1.4233 to 1.4253
25 g
1-amino-3,3-diethoxypropane, 3,3-diethoxypropylamine, 3-aminopropionaldehyde diethylacetal, 1-propanamine, 3,3-diethoxy, 3-aminopropionaldehyde diethyl acetal, propionaldehyde, 3-amino-, diethyl acetal, zlchem 1268, acmc-1apws, 3,3-diethoxy-propylamine, 3,3-diethoxypropyl amine
PXXMSHBZYAOHBD-UHFFFAOYSA-N
3,3-diethoxypropan-1-amine
521029
97%
1-Amino-3, 3-diethoxypropane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only