missing translation for 'onlineSavingsMsg'
Learn More

1-Allylpiperazine, 98+%

CAS: 13961-36-9 | C7H14N2 | 126.203 g/mol

$156.75 - $545.48

Chemical Identifiers

CAS 13961-36-9
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.203
MDL Number MFCD00167970
InChI Key ZWAQJGHGPPDZSF-UHFFFAOYSA-N
Synonym 1-allylpiperazine, 1-allyl-piperazine, 1-allyl piperazine, 1-prop-2-en-1-yl piperazine, n-allyl piperazine, piperazine,1-2-propen-1-yl, prop-2-enylpiperazine, allylpiperazin, allylpiperazine, 1-allylpiperazin
PubChem CID 806422
IUPAC Name 1-prop-2-enylpiperazine
SMILES C=CCN1CCNCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1939106
View Documents
Thermo Scientific Chemicals
L1939106
5 g
Each for $156.75
Only null left
 
AAL1939114
View Documents
Thermo Scientific Chemicals
L1939114
25 g
Each for $545.48
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13961-36-9
126.203
ZWAQJGHGPPDZSF-UHFFFAOYSA-N
806422
C=CCN1CCNCC1
C7H14N2
MFCD00167970
1-allylpiperazine, 1-allyl-piperazine, 1-allyl piperazine, 1-prop-2-en-1-yl piperazine, n-allyl piperazine, piperazine,1-2-propen-1-yl, prop-2-enylpiperazine, allylpiperazin, allylpiperazine, 1-allylpiperazin
1-prop-2-enylpiperazine

Specifications

13961-36-9
180°C to 182°C
C7H14N2
MFCD00167970
UN2920
Air Sensitive
ZWAQJGHGPPDZSF-UHFFFAOYSA-N
1-prop-2-enylpiperazine
806422
≥98%
0.902
51°C (123°F)
1.483
5 g
507447
1-allylpiperazine, 1-allyl-piperazine, 1-allyl piperazine, 1-prop-2-en-1-yl piperazine, n-allyl piperazine, piperazine,1-2-propen-1-yl, prop-2-enylpiperazine, allylpiperazin, allylpiperazine, 1-allylpiperazin
C=CCN1CCNCC1
126.203
126.2
1-Allylpiperazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H332-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only