missing translation for 'onlineSavingsMsg'
Learn More

1-Allylpiperazine, 98+%

Catalog No. AAL1939106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1939106 5 g
AAL1939114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1939106 Supplier Thermo Scientific Chemicals Supplier No. L1939106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13961-36-9 | C7H14N2 | 126.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13961-36-9
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.203
MDL Number MFCD00167970
InChI Key ZWAQJGHGPPDZSF-UHFFFAOYSA-N
Synonym 1-allylpiperazine, 1-allyl-piperazine, 1-allyl piperazine, 1-prop-2-en-1-yl piperazine, n-allyl piperazine, piperazine,1-2-propen-1-yl, prop-2-enylpiperazine, allylpiperazin, allylpiperazine, 1-allylpiperazin
PubChem CID 806422
IUPAC Name 1-prop-2-enylpiperazine
SMILES C=CCN1CCNCC1

Specifications

Density 0.902
Boiling Point 180°C to 182°C
Flash Point 51°C (123°F)
Refractive Index 1.483
Quantity 5 g
UN Number UN2920
Beilstein 507447
Sensitivity Air Sensitive
Formula Weight 126.2
Percent Purity ≥98%
Chemical Name or Material 1-Allylpiperazine
Show More Show Less
Hazard Category H226-H314-H332-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only