missing translation for 'onlineSavingsMsg'
Learn More

1-Allylimidazole 97.0+%, TCI America™

Catalog No. A220725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A220725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A220725G Supplier TCI America Supplier No. A220725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31410-01-2
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00037894
InChI Key XLXCHZCQTCBUOX-UHFFFAOYSA-N
Synonym 1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole #
PubChem CID 35794
IUPAC Name 1-(prop-2-en-1-yl)-1H-imidazole
SMILES C=CCN1C=CN=C1

Specifications

Color Yellow
Boiling Point 116°C
Quantity 25 g
UN Number 3267
Formula Weight 108.14
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Allylimidazole