missing translation for 'onlineSavingsMsg'
Learn More

1-Allylimidazole, 97%

Catalog No. AAL1105318
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1105318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1105318 Supplier Thermo Scientific Chemicals Supplier No. L1105318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31410-01-2 | C6H8N2 | 108.14 g/mol

1-Allylimidazole, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Allylimidazole, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 31410-01-2
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00037894
InChI Key XLXCHZCQTCBUOX-UHFFFAOYSA-N
Synonym 1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole #
PubChem CID 35794
IUPAC Name 1-prop-2-enylimidazole
SMILES C=CCN1C=CN=C1

Specifications

Density 1.003
Boiling Point 212°C to 213°C
Flash Point 101°C (213°F)
Refractive Index 1.505
Quantity 50 g
UN Number UN3267
Beilstein 605953
Solubility Information It is soluble in water.
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 1-Allylimidazole
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 250-618-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only