missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate 96.0+%, TCI America™
Supplier: TCI America A29095G
Specifications
| 1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate | |
| 118°C | |
| C23H23NO6 | |
| 5 g | |
| ORKKMGRINLTBPC-UHFFFAOYNA-N | |
| 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid | |
| 7020606 | |
| ≥96.0% (HPLC,T) |
| 144120-54-7 | |
| White-Yellow | |
| MFCD00467718 | |
| fmoc-glu-oall, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-allyl ester, fmoc-l-glutamic acid 1-allyl ester, fmoc-l-glutamic acid, fmoc-l-glutamic acid, fmoc-glu-allyl ester, fmoc-glu-allyl ester, n-fmoc-l-glutamic acid 1-allyl ester, n-fmoc-l-glutamic acid 1-allyl ester, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate | |
| OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC=C | |
| 409.44 | |
| 409.44 | |
| Crystalline Powder |
Chemical Identifiers
| 144120-54-7 | |
| 409.44 | |
| ORKKMGRINLTBPC-UHFFFAOYNA-N | |
| 7020606 | |
| OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC=C |
| C23H23NO6 | |
| MFCD00467718 | |
| fmoc-glu-oall, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-allyl ester, fmoc-l-glutamic acid 1-allyl ester, fmoc-l-glutamic acid, fmoc-l-glutamic acid, fmoc-glu-allyl ester, fmoc-glu-allyl ester, n-fmoc-l-glutamic acid 1-allyl ester, n-fmoc-l-glutamic acid 1-allyl ester, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate | |
| 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator