missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Allyl-3-methylimidazolium Chloride 98.0+%, TCI America™
Supplier: TCI America A20845G
Specifications
| 1-Allyl-3-methylimidazolium Chloride | |
| C7H11ClN2 | |
| 5 g | |
| QVRCRKLLQYOIKY-UHFFFAOYSA-M | |
| 1-methyl-3-prop-2-enylimidazol-1-ium;chloride | |
| 11321106 | |
| ≥98.0% (HPLC,T) |
| 65039-10-3 | |
| MFCD07784450 | |
| 1-allyl-3-methylimidazolium chloride, 1-allyl-3-methyl-1h-imidazol-3-ium chloride, 1-methyl-3-prop-2-en-1-yl imidazol-1-ium chloride, amim-cl, 1-allyl-3-methylimidazoliumchloride, dsstox_cid_27904, dsstox_rid_82657, dsstox_gsid_47928, 1-methyl-3-allylimidazolium chloride, 1-allyl-3-methyl-3-imidazolium chloride | |
| C[N+]1=CN(C=C1)CC=C.[Cl-] | |
| 158.629 | |
| 158.63 | |
| Liquid |
Chemical Identifiers
| 65039-10-3 | |
| 158.629 | |
| QVRCRKLLQYOIKY-UHFFFAOYSA-M | |
| 11321106 | |
| C[N+]1=CN(C=C1)CC=C.[Cl-] |
| C7H11ClN2 | |
| MFCD07784450 | |
| 1-allyl-3-methylimidazolium chloride, 1-allyl-3-methyl-1h-imidazol-3-ium chloride, 1-methyl-3-prop-2-en-1-yl imidazol-1-ium chloride, amim-cl, 1-allyl-3-methylimidazoliumchloride, dsstox_cid_27904, dsstox_rid_82657, dsstox_gsid_47928, 1-methyl-3-allylimidazolium chloride, 1-allyl-3-methyl-3-imidazolium chloride | |
| 1-methyl-3-prop-2-enylimidazol-1-ium;chloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator