missing translation for 'onlineSavingsMsg'
Learn More

1-Allyl-3-methylimidazolium chloride, 98%

Catalog No. AAH2695206
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2695206 5 g
AAH2695218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2695206 Supplier Thermo Scientific Chemicals Supplier No. H2695206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65039-10-3 | C7H11ClN2 | 158.629 g/mol

1-Allyl-3-methylimidazolium chloride is used as an ionic liquid as well as nonderivatizing solvent for cellulose. It acts as a plasticizer for cornstarch and finds application as a solid biopolymer electrolyte. Further, it plays an important role in the preparation of reducing sugar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Allyl-3-methylimidazolium chloride is used as an ionic liquid as well as nonderivatizing solvent for cellulose. It acts as a plasticizer for cornstarch and finds application as a solid biopolymer electrolyte. Further, it plays an important role in the preparation of reducing sugar.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and bases.

Chemical Identifiers

CAS 65039-10-3
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.629
MDL Number MFCD07784450
InChI Key QVRCRKLLQYOIKY-UHFFFAOYSA-M
Synonym 1-allyl-3-methylimidazolium chloride, 1-allyl-3-methyl-1h-imidazol-3-ium chloride, 1-methyl-3-prop-2-en-1-yl imidazol-1-ium chloride, amim-cl, 1-allyl-3-methylimidazoliumchloride, dsstox_cid_27904, dsstox_rid_82657, dsstox_gsid_47928, 1-methyl-3-allylimidazolium chloride, 1-allyl-3-methyl-3-imidazolium chloride
PubChem CID 11321106
IUPAC Name 1-methyl-3-prop-2-enylimidazol-1-ium;chloride
SMILES C[N+]1=CN(C=C1)CC=C.[Cl-]

Specifications

Melting Point 49°C to 51°C
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 158.63
Percent Purity 98%
Chemical Name or Material 1-Allyl-3-methylimidazolium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only