missing translation for 'onlineSavingsMsg'
Learn More

1-Allyl-1-methylpyrrolidinium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. A30895G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A30895G Supplier TCI America Supplier No. A30895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1059624-23-5
Molecular Formula C10H16F6N2O4S2
Molecular Weight (g/mol) 406.358
InChI Key RXYDHDCXRMFQBC-UHFFFAOYSA-N
PubChem CID 132274870
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-methyl-1-prop-2-enylpyrrolidin-1-ium
SMILES C[N+]1(CCCC1)CC=C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 406.36
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Allyl-1-methylpyrrolidinium Bis(trifluoromethanesulfonyl)imide