missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantyl methyl ketone, 99%

Catalog No. AC164740250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164740250 25 g Glass bottle
AC164740050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC164740250 Supplier Thermo Scientific Chemicals Supplier No. 164740250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1660-04-4 | C12H18O | 178.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1660-04-4
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD00074739
InChI Key DACIGVIOAFXPHW-UHFFFAOYSA-N
Synonym 1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone
PubChem CID 123126
IUPAC Name 1-(1-adamantyl)ethanone
SMILES CC(=O)C12CC3CC(CC(C3)C1)C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53°C to 55°C
Color White
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Formula Weight 178.27
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantyl methyl ketone, 99%
Show More Show Less
EINECSNumber 216-761-9
TSCA TSCA

RUO – Research Use Only