missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantyl Isocyanate 98.0+%, TCI America™

Catalog No. A17061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A17061G 1 g
A17065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A17061G Supplier TCI America Supplier No. A17061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4411-25-0
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.25
MDL Number MFCD00134405
InChI Key VBHCPGFCIQDXGZ-UHFFFAOYSA-N
Synonym 1-adamantyl isocyanate, 1-isocyanato-adamantane, 1-isocyantoadamantane, 1-adamantylisocyanate, adamantanisocyanate, adamantyl isocyanate, pubchem8848, 1-adamantyl-isocyanate, 1-adamantanyl isocyanate, adamantane, 1-isocyanato
PubChem CID 2059995
IUPAC Name 1-isocyanatoadamantane
SMILES O=C=NC12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 147°C
Color White
Quantity 1 g
Formula Weight 177.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantyl Isocyanate