Learn More
1-Adamantyl isocyanate, 98%
CAS: 4411-25-0 | C11H15NO | 177.25 g/mol
Supplier: Thermo Scientific Chemicals 314360050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Adamantyl isocyanate | |
| 97.5 | |
| 144.0°C to 146.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00134405 | |
| 1-adamantyl isocyanate, 1-isocyanato-adamantane, 1-isocyantoadamantane, 1-adamantylisocyanate, adamantanisocyanate, adamantyl isocyanate, pubchem8848, 1-adamantyl-isocyanate, 1-adamantanyl isocyanate, adamantane, 1-isocyanato | |
| O=C=NC12CC3CC(CC(C3)C1)C2 | |
| 177.25 | |
| 177.25 | |
| Powder |
| 4411-25-0 | |
| 100.0 | |
| White | |
| 97.5% min. (GC) | |
| C11H15NO | |
| 5 g | |
| VBHCPGFCIQDXGZ-UHFFFAOYSA-N | |
| 1-isocyanatoadamantane | |
| 2059995 | |
| 98% |
Chemical Identifiers
| 4411-25-0 | |
| 177.25 | |
| VBHCPGFCIQDXGZ-UHFFFAOYSA-N | |
| 2059995 | |
| O=C=NC12CC3CC(CC(C3)C1)C2 |
| C11H15NO | |
| MFCD00134405 | |
| 1-adamantyl isocyanate, 1-isocyanato-adamantane, 1-isocyantoadamantane, 1-adamantylisocyanate, adamantanisocyanate, adamantyl isocyanate, pubchem8848, 1-adamantyl-isocyanate, 1-adamantanyl isocyanate, adamantane, 1-isocyanato | |
| 1-isocyanatoadamantane |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
RUO – Research Use Only