missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Adamantanol 98.0+%, TCI America™
$143.54 - $719.52
Chemical Identifiers
| CAS | 768-95-6 |
|---|---|
| Molecular Formula | C10H16O |
| Molecular Weight (g/mol) | 152.237 |
| MDL Number | MFCD00074729 |
| InChI Key | VLLNJDMHDJRNFK-UHFFFAOYSA-N |
| Synonym | 1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol |
| PubChem CID | 64152 |
| IUPAC Name | adamantan-1-ol |
| SMILES | C1C2CC3CC1CC(C2)(C3)O |
Chemical Identifiers
| 768-95-6 | |
| 152.237 | |
| VLLNJDMHDJRNFK-UHFFFAOYSA-N | |
| 64152 | |
| C1C2CC3CC1CC(C2)(C3)O |
| C10H16O | |
| MFCD00074729 | |
| 1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol | |
| adamantan-1-ol |
Specifications
| 768-95-6 | |
| White | |
| MFCD00074729 | |
| 1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol | |
| C1C2CC3CC1CC(C2)(C3)O | |
| 152.237 | |
| 152.24 | |
| Crystalline Powder |
| 280°C | |
| C10H16O | |
| 25 g | |
| VLLNJDMHDJRNFK-UHFFFAOYSA-N | |
| adamantan-1-ol | |
| 64152 | |
| ≥98.0% (GC) | |
| 1-Adamantanol |
Safety and Handling
EINECSNumber : (4)-1945
RTECSNumber : AU4980000
TSCA : No