missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanol, 99%

Catalog No. AC157100250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157100250 25 g Glass bottle
AC157101000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC157100250 Supplier Thermo Scientific Chemicals Supplier No. 157100250

CAS: 768-95-6 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 768-95-6
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00074729
InChI Key VLLNJDMHDJRNFK-UHFFFAOYSA-N
Synonym 1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol
PubChem CID 64152
IUPAC Name adamantan-1-ol
SMILES C1C2CC3CC1CC(C2)(C3)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >240.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol,diethyl ether,methylene,chloride and acetone
Formula Weight 152.24
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 1-Adamantanol
Show More Show Less
EINECSNumber 212-202-8

RUO – Research Use Only