missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Adamantanol, 99%
CAS: 768-95-6 | C10H16O | 152.237 g/mol
$73.62 - $1,401.66
Chemical Identifiers
| CAS | 768-95-6 |
|---|---|
| Molecular Formula | C10H16O |
| Molecular Weight (g/mol) | 152.237 |
| MDL Number | MFCD00074729 |
| InChI Key | VLLNJDMHDJRNFK-UHFFFAOYSA-N |
| Synonym | 1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol |
| PubChem CID | 64152 |
| IUPAC Name | adamantan-1-ol |
| SMILES | C1C2CC3CC1CC(C2)(C3)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1020909
|
Thermo Scientific Chemicals
A1020909 |
10 g |
N/A
|
|
|||||
|
AAA1020918
|
Thermo Scientific Chemicals
A1020918 |
50 g |
N/A
|
|
|||||
|
AAA1020930
|
Thermo Scientific Chemicals
A1020930 |
250 g |
N/A
|
|
|||||
Description
1-Adamantanol was used in the preparation of 1,3-adamantanediol. It is used in the manufacture of synthetic adamantane derivatives and adapalene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Adamantanol was used in the preparation of 1,3-adamantanediol. It is used in the manufacture of synthetic adamantane derivatives and adapalene.
Solubility
Soluble in ethanol , methanol, and diethyl ether. Partly miscible in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
Chemical Identifiers
| 768-95-6 | |
| 152.237 | |
| VLLNJDMHDJRNFK-UHFFFAOYSA-N | |
| 64152 | |
| C1C2CC3CC1CC(C2)(C3)O |
| C10H16O | |
| MFCD00074729 | |
| 1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol | |
| adamantan-1-ol |
Specifications
| 768-95-6 | |
| C10H16O | |
| 10 g | |
| 1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol | |
| VLLNJDMHDJRNFK-UHFFFAOYSA-N | |
| adamantan-1-ol | |
| 64152 | |
| 99% |
| ∼245°C (sublimation) | |
| MFCD00074729 | |
| 1903952 | |
| Soluble in ethanol ,methanol,and diethyl ether. Partly miscible in water. | |
| C1C2CC3CC1CC(C2)(C3)O | |
| 152.237 | |
| 152.24 | |
| 1-Adamantanol |
Safety and Handling
EINECSNumber : 212-202-8
RTECSNumber : AU4980000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only