missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanemethanol 99.0+%, TCI America™

Catalog No. p-7130534
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04201G 1 g
H04205G 5 g
H042025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H04201G Supplier TCI America Supplier No. H04201G

Chemical Identifiers

CAS 770-71-8
Molecular Formula C11H18O
Molecular Weight (g/mol) 166.264
MDL Number MFCD00074751
InChI Key MDVGOOIANLZFCP-UHFFFAOYSA-N
Synonym 1-adamantanemethanol, adamantan-1-ylmethanol, 1-adamantane methanol, 1-hydroxymethyl adamantane, adamantane-1-methanol, adamantan-1-yl-methanol, 1-hydroxymethyladamantane, tricyclo 3.3.1.1'3,7 dec-1-ylmethanol, adamantan-1-yl methanol, tricyclo 3.3.1.13,7 decanemethanol
PubChem CID 64556
IUPAC Name 1-adamantylmethanol
SMILES C1C2CC3CC1CC(C2)(C3)CO

Specifications

Melting Point 118°C
Color White
Quantity 1 g
Formula Weight 166.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantanemethanol