missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanemethanol, 99%

Catalog No. p-50427
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC180050100 10 g Glass bottle
AC180050500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC180050100 Supplier Thermo Scientific Chemicals Supplier No. 180050100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 770-71-8 | C11H18O | 166.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 770-71-8
Molecular Formula C11H18O
Molecular Weight (g/mol) 166.26
MDL Number MFCD00074751
InChI Key MDVGOOIANLZFCP-UHFFFAOYSA-N
Synonym 1-adamantanemethanol, adamantan-1-ylmethanol, 1-adamantane methanol, 1-hydroxymethyl adamantane, adamantane-1-methanol, adamantan-1-yl-methanol, 1-hydroxymethyladamantane, tricyclo 3.3.1.1'3,7 dec-1-ylmethanol, adamantan-1-yl methanol, tricyclo 3.3.1.13,7 decanemethanol
PubChem CID 64556
IUPAC Name 1-adamantylmethanol
SMILES C1C2CC3CC1CC(C2)(C3)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 115.0°C to 118.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: .. Other solubilities: soluble in hexane and ethyl alcohol
Formula Weight 166.26
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantanemethanol, 99%
Show More Show Less
EINECSNumber 212-225-3
TSCA TSCA

RUO – Research Use Only