missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7117038
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A074225G 25 g
A0742100G 100 g
A0742500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A074225G Supplier TCI America Supplier No. A074225G
Only null left

Chemical Identifiers

CAS 828-51-3
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.247
MDL Number MFCD00074720
InChI Key JIMXXGFJRDUSRO-UHFFFAOYSA-N
Synonym 1-adamantanecarboxylic acid, 1-carboxyadamantane, adamantanecarboxylic acid, adamantoic acid, 1-adamantoic acid, 3-adamantanecarboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, tricyclo 3.3.1.1'3,7 decane-1-carboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3r,5s,7s-adamantane-1-carboxylic acid
PubChem CID 13235
IUPAC Name adamantane-1-carboxylic acid
SMILES C1C2CC3CC1CC(C2)(C3)C(=O)O

Specifications

Melting Point 173°C
Color White
Quantity 25 g
Formula Weight 180.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantanecarboxylic Acid
RTECSNumber AU4452200
TSCA No