missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantaneacetic Acid 99.0+%, TCI America™

Catalog No. A07365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07365G 5 g
A073625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A07365G Supplier TCI America Supplier No. A07365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4942-47-6
Molecular Formula C12H17O2
Molecular Weight (g/mol) 193.27
MDL Number MFCD00074728
InChI Key AOTQGWFNFTVXNQ-UHFFFAOYSA-M
Synonym 1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane
PubChem CID 123221
IUPAC Name 2-(adamantan-1-yl)acetate
SMILES [O-]C(=O)CC12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 137°C
Quantity 5 g
Formula Weight 194.27
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantaneacetic Acid