missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantaneacetic acid, 98%

CAS: 4942-47-6 | C12H17O2 | 193.27 g/mol

$73.56 - $73.56

Chemical Identifiers

CAS 4942-47-6
Molecular Formula C12H17O2
Molecular Weight (g/mol) 193.27
MDL Number MFCD00074728
InChI Key AOTQGWFNFTVXNQ-UHFFFAOYSA-M
Synonym 1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane
PubChem CID 123221
IUPAC Name 2-(1-adamantyl)acetic acid
SMILES [O-]C(=O)CC12CC3CC(CC(C3)C1)C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC331560010
SDS View Documents
Thermo Scientific Chemicals
331560010
1 g Glass bottle
Each for $73.56
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4942-47-6
193.27
AOTQGWFNFTVXNQ-UHFFFAOYSA-M
123221
[O-]C(=O)CC12CC3CC(CC(C3)C1)C2
C12H17O2
MFCD00074728
1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane
2-(1-adamantyl)acetic acid

Specifications

4942-47-6
100.0
White
98%
C12H17O2
1 g
AOTQGWFNFTVXNQ-UHFFFAOYSA-M
2-(1-adamantyl)acetic acid
123221
98%
1-Adamantaneacetic acid
97.5
138°C to 140°C
Authentic
Glass bottle
MFCD00074728
1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane
[O-]C(=O)CC12CC3CC(CC(C3)C1)C2
193.27
194.27
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 225-585-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only