Learn More
1-Adamantaneacetic acid, 98+%
CAS: 4942-47-6 | C12H17O2 | 193.27 g/mol
Supplier: Thermo Scientific Chemicals L1202906
Description
1-Adamantaneacetic acid is used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Adamantaneacetic acid is used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.
Solubility
Solubility in Methanol - almost transparency
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place
Specifications
| 1-Adamantaneacetic acid | |
| 136°C to 138°C | |
| MFCD00074728 | |
| 641412 | |
| AOTQGWFNFTVXNQ-UHFFFAOYSA-M | |
| 2-(1-adamantyl)acetic acid | |
| 123221 | |
| ≥98% |
| 4942-47-6 | |
| C12H17O2 | |
| 5 g | |
| 1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane | |
| [O-]C(=O)CC12CC3CC(CC(C3)C1)C2 | |
| 193.27 | |
| 194.28 |
Chemical Identifiers
| 4942-47-6 | |
| 193.27 | |
| AOTQGWFNFTVXNQ-UHFFFAOYSA-M | |
| 123221 | |
| [O-]C(=O)CC12CC3CC(CC(C3)C1)C2 |
| C12H17O2 | |
| MFCD00074728 | |
| 1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane | |
| 2-(1-adamantyl)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 225-585-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only