missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantaneacetic acid, 98+%

Catalog No. AAL1202914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1202914 25 g
AAL1202903 1 g
AAL1202906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1202914 Supplier Thermo Scientific Chemicals Supplier No. L1202914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4942-47-6 | C12H17O2 | 193.27 g/mol

1-Adamantaneacetic acid is used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantaneacetic acid is used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.

Solubility
Solubility in Methanol - almost transparency

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place

Chemical Identifiers

CAS 4942-47-6
Molecular Formula C12H17O2
Molecular Weight (g/mol) 193.27
MDL Number MFCD00074728
InChI Key AOTQGWFNFTVXNQ-UHFFFAOYSA-M
Synonym 1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane
PubChem CID 123221
IUPAC Name 2-(1-adamantyl)acetic acid
SMILES [O-]C(=O)CC12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 136°C to 138°C
Quantity 25 g
Beilstein 641412
Formula Weight 194.28
Percent Purity ≥98%
Chemical Name or Material 1-Adamantaneacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-585-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only