missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanamine, 98%

Catalog No. p-7030530
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3007606 5 g
AAH3007614 25 g
AAH3007622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3007606 Supplier Thermo Scientific Chemicals Supplier No. H3007606

CAS: 768-94-5 | C10H17N | 151.253 g/mol

1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Use only in a chemical fume hood. Keep away from sources of ignition. Incompatible with oxidizing agent.

Chemical Identifiers

CAS 768-94-5
Molecular Formula C10H17N
Molecular Weight (g/mol) 151.253
MDL Number MFCD00074732
InChI Key DKNWSYNQZKUICI-UHFFFAOYSA-N
Synonym amantadine, 1-adamantanamine, 1-adamantylamine, 1-aminoadamantane, adamantanamine, adamantylamine, aminoadamantane, amantidine, symadine, symmetrel
PubChem CID 2130
ChEBI CHEBI:2618
IUPAC Name adamantan-1-amine
SMILES C1C2CC3CC1CC(C2)(C3)N

Specifications

Melting Point ∼200°C (sublimation)
Quantity 5 g
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 151.25
Percent Purity 98%
Chemical Name or Material 1-Adamantanamine
Hazard Category Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, Causes skin irritation
EINECSNumber 212-201-2
RTECSNumber YD1925000
TSCA Yes

RUO – Research Use Only