missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpyrrolidine 97.0+%, TCI America™

Catalog No. A10305G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10305G Supplier TCI America Supplier No. A10305G
Only null left

Chemical Identifiers

CAS 4030-18-6
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00014096
InChI Key LNWWQYYLZVZXKS-UHFFFAOYSA-N
PubChem CID 77650
IUPAC Name 1-pyrrolidin-1-ylethanone
SMILES CC(=O)N1CCCC1

Specifications

Color Yellow
Boiling Point 225°C
Quantity 5 g
Formula Weight 113.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Acetylpyrrolidine