missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpiperidine-4-carbonyl chloride, 97%, may contain up to ca 1M free HCl

Catalog No. AAL1842606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1842606 5 g
AAL1842603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1842606 Supplier Thermo Scientific Chemicals Supplier No. L1842606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 59084-16-1 | C8H12ClNO2 | 189.639 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59084-16-1
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.639
MDL Number MFCD02094017
InChI Key OHCPVLJEAHBMEG-UHFFFAOYSA-N
Synonym 1-acetylisonipecotoyl chloride, n-acetylisonipecotoyl chloride, 1-acetyl-4-piperidinecarbonyl chloride, 1-acetyl isonipecotoyl chloride, acmc-20aoys, acetylisonipecotoyl chloride, 1acetylisonipecotoyl chloride, n-acetylisonipecotyl chloride, ksc274a6b, n-acetyl isonipecotoyl chloride
PubChem CID 100950
IUPAC Name 1-acetylpiperidine-4-carbonyl chloride
SMILES CC(=O)N1CCC(CC1)C(=O)Cl

Specifications

Melting Point 131°C to 133°C
Quantity 5 g
UN Number UN3261
Beilstein 389571
Sensitivity Moisture sensitive
Formula Weight 189.64
Percent Purity 97%
Chemical Name or Material 1-Acetylpiperidine-4-carbonyl chloride, may contain up to ≈1M free HCl
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 261-594-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only