missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acetylpiperazine 98.0+%, TCI America™
$32.72 - $73.16
Chemical Identifiers
| CAS | 13889-98-0 |
|---|---|
| Molecular Formula | C6H12N2O |
| Molecular Weight (g/mol) | 128.175 |
| MDL Number | MFCD00058676 |
| InChI Key | PKDPUENCROCRCH-UHFFFAOYSA-N |
| Synonym | 1-acetylpiperazine, n-acetylpiperazine, 1-piperazin-1-yl ethanone, 1-piperazin-1-yl ethan-1-one, n-acetyl piperazine, acetylpiperazine, 1-acetyl piperazine, piperazine, 1-acetyl, 1-piperazin-1-yl-ethanone, 1-piperazin-1-yl ethanon |
| PubChem CID | 83795 |
| IUPAC Name | 1-piperazin-1-ylethanone |
| SMILES | CC(=O)N1CCNCC1 |
Chemical Identifiers
| 13889-98-0 | |
| 128.175 | |
| PKDPUENCROCRCH-UHFFFAOYSA-N | |
| 83795 | |
| CC(=O)N1CCNCC1 |
| C6H12N2O | |
| MFCD00058676 | |
| 1-acetylpiperazine, n-acetylpiperazine, 1-piperazin-1-yl ethanone, 1-piperazin-1-yl ethan-1-one, n-acetyl piperazine, acetylpiperazine, 1-acetyl piperazine, piperazine, 1-acetyl, 1-piperazin-1-yl-ethanone, 1-piperazin-1-yl ethanon | |
| 1-piperazin-1-ylethanone |
Specifications
| 13889-98-0 | |
| C6H12N2O | |
| 5 g | |
| PKDPUENCROCRCH-UHFFFAOYSA-N | |
| 1-piperazin-1-ylethanone | |
| 83795 | |
| ≥98.0% (GC,T) | |
| 1-Acetylpiperazine |
| White-Yellow | |
| MFCD00058676 | |
| 1-acetylpiperazine, n-acetylpiperazine, 1-piperazin-1-yl ethanone, 1-piperazin-1-yl ethan-1-one, n-acetyl piperazine, acetylpiperazine, 1-acetyl piperazine, piperazine, 1-acetyl, 1-piperazin-1-yl-ethanone, 1-piperazin-1-yl ethanon | |
| CC(=O)N1CCNCC1 | |
| 128.175 | |
| 128.18 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No