missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpiperazine 98.0+%, TCI America™

Catalog No. A13995G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13995G 5 g
A139925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. A13995G missing translation for 'mfr' TCI America missing translation for 'supplierNo' A13995G

Chemical Identifiers

CAS 13889-98-0
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.175
MDL Number MFCD00058676
InChI Key PKDPUENCROCRCH-UHFFFAOYSA-N
Synonym 1-acetylpiperazine, n-acetylpiperazine, 1-piperazin-1-yl ethanone, 1-piperazin-1-yl ethan-1-one, n-acetyl piperazine, acetylpiperazine, 1-acetyl piperazine, piperazine, 1-acetyl, 1-piperazin-1-yl-ethanone, 1-piperazin-1-yl ethanon
PubChem CID 83795
IUPAC Name 1-piperazin-1-ylethanone
SMILES CC(=O)N1CCNCC1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 128.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Acetylpiperazine