missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpiperazine, 98%

CAS: 13889-98-0 | C6H12N2O | 128.175 g/mol

Supplier:  Thermo Scientific Chemicals B2020022

 View more versions of this product

Catalog No. AAB2020022


Only null left
Explore available promotions

Description

Description

1-Acetylpiperazine may be used in the synthesis of series of 7-alkoxyl substituted indolizinoquinoline-5,12-dione derivatives and 2-substituted-N-(naphth-1-ylmethyl)-pyrimidin-4-amines and 2-substituted-N-benzhydrylpyrimidin-4-amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Acetylpiperazine may be used in the synthesis of series of 7-alkoxyl substituted indolizinoquinoline-5,12-dione derivatives and 2-substituted-N-(naphth-1-ylmethyl)-pyrimidin-4-amines and 2-substituted-N-benzhydrylpyrimidin-4-amines.

Solubility
Soluble in water 210 g/L (20°C).

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
Specifications

Specifications

1-Acetylpiperazine
30°C to 34°C
>110°C (230°F)
MFCD00058676
112220
1-acetylpiperazine, n-acetylpiperazine, 1-piperazin-1-yl ethanone, 1-piperazin-1-yl ethan-1-one, n-acetyl piperazine, acetylpiperazine, 1-acetyl piperazine, piperazine, 1-acetyl, 1-piperazin-1-yl-ethanone, 1-piperazin-1-yl ethanon
PKDPUENCROCRCH-UHFFFAOYSA-N
1-piperazin-1-ylethanone
83795
99%
13889-98-0
127°C to 130°C (8 mmHg)
C6H12N2O
100 g
Hygroscopic
Soluble in water 210g/L (20°C).
CC(=O)N1CCNCC1
128.175
128.18

Chemical Identifiers

13889-98-0
1-acetylpiperazine, n-acetylpiperazine, 1-piperazin-1-yl ethanone, 1-piperazin-1-yl ethan-1-one, n-acetyl piperazine, acetylpiperazine, 1-acetyl piperazine, piperazine, 1-acetyl, 1-piperazin-1-yl-ethanone, 1-piperazin-1-yl ethanon
1-piperazin-1-ylethanone
MFCD00058676
83795
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-659-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only