missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpiperazine, 98%

Catalog No. AAB2020006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2020006 5 g
AAB2020014 25 g
AAB2020022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2020006 Supplier Thermo Scientific Chemicals Supplier No. B2020006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13889-98-0 | C6H12N2O | 128.175 g/mol

1-Acetylpiperazine may be used in the synthesis of series of 7-alkoxyl substituted indolizinoquinoline-5,12-dione derivatives and 2-substituted-N-(naphth-1-ylmethyl)-pyrimidin-4-amines and 2-substituted-N-benzhydrylpyrimidin-4-amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Acetylpiperazine may be used in the synthesis of series of 7-alkoxyl substituted indolizinoquinoline-5,12-dione derivatives and 2-substituted-N-(naphth-1-ylmethyl)-pyrimidin-4-amines and 2-substituted-N-benzhydrylpyrimidin-4-amines.

Solubility
Soluble in water 210 g/L (20°C).

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.

Chemical Identifiers

CAS 13889-98-0
MDL Number MFCD00058676
Synonym 1-acetylpiperazine, n-acetylpiperazine, 1-piperazin-1-yl ethanone, 1-piperazin-1-yl ethan-1-one, n-acetyl piperazine, acetylpiperazine, 1-acetyl piperazine, piperazine, 1-acetyl, 1-piperazin-1-yl-ethanone, 1-piperazin-1-yl ethanon
PubChem CID 83795
IUPAC Name 1-piperazin-1-ylethanone

Specifications

Melting Point 30°C to 34°C
Boiling Point 127°C to 130°C (8 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C6H12N2O
Quantity 5 g
Beilstein 112220
Sensitivity Hygroscopic
Solubility Information Soluble in water 210g/L (20°C).
InChI Key PKDPUENCROCRCH-UHFFFAOYSA-N
SMILES CC(=O)N1CCNCC1
Molecular Weight (g/mol) 128.175
Formula Weight 128.18
Percent Purity 99%
Chemical Name or Material 1-Acetylpiperazine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-659-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only