missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylisatin 98.0+%, TCI America™

Catalog No. a294125g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A294125G 25 g
A29415G 5 g
2 options

Catalog No. A294125G

Supplier: TCI America A294125G

Chemical Identifiers

CAS 574-17-4
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00158542
InChI Key LPGDEHBASRKTDG-UHFFFAOYSA-N
Synonym 1-acetylisatin, n-acetylisatin, 1-acetyl-1h-indole-2,3-dione, acetylisatin, 1-acetylindoline-2,3-dione, 1-acetyl-indole-2,3-dione, 1h-indole-2,3-dione, 1-acetyl, indole-2,3-dione, 1-acetyl, 1-acetyl-2,3-dihydro-1h-indole-2,3-dione, n-acetyl-isatin
PubChem CID 11321
ChEBI CHEBI:16050
IUPAC Name 1-acetyl-2,3-dihydro-1H-indole-2,3-dione
SMILES CC(=O)N1C(=O)C(=O)C2=CC=CC=C12

Specifications

Melting Point 143°C
Quantity 25 g
Formula Weight 189.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetylisatin
RTECSNumber NL7873800
TSCA No