missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylisatin, 97%

Catalog No. p-7047097
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1759703 1 g
AAL1759706 5 g
AAL1759714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1759703 Supplier Thermo Scientific Chemicals Supplier No. L1759703

CAS: 574-17-4 | C10H7NO3 | 189.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 574-17-4
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00158542
InChI Key LPGDEHBASRKTDG-UHFFFAOYSA-N
Synonym 1-acetylisatin, n-acetylisatin, 1-acetyl-1h-indole-2,3-dione, acetylisatin, 1-acetylindoline-2,3-dione, 1-acetyl-indole-2,3-dione, 1h-indole-2,3-dione, 1-acetyl, indole-2,3-dione, 1-acetyl, 1-acetyl-2,3-dihydro-1h-indole-2,3-dione, n-acetyl-isatin
PubChem CID 11321
ChEBI CHEBI:16050
IUPAC Name 1-acetylindole-2,3-dione
SMILES CC(=O)N1C(=O)C(=O)C2=CC=CC=C12

Specifications

Melting Point 141°C to 143°C
Quantity 1 g
Beilstein 113959
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 1-Acetylisatin
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-368-9
RTECSNumber NL7873800
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only