Learn More
1-Acetylindoline-5-sulfonyl chloride, 97%
CAS: 52206-05-0 | C10H10ClNO3S | 259.70 g/mol
$155.70 - $737.90
Chemical Identifiers
| CAS | 52206-05-0 |
|---|---|
| Molecular Formula | C10H10ClNO3S |
| Molecular Weight (g/mol) | 259.70 |
| MDL Number | MFCD07368558 |
| InChI Key | QNFXLCHANYHGIF-UHFFFAOYSA-N |
| Synonym | 1-acetyl-2,3-dihydro-1h-indole-5-sulfonyl chloride, 1-acetylindoline-5-sulfonyl chloride, 1-acetyl-5-indolinesulfonoyl chloride, 1h-indole-5-sulfonyl chloride, 1-acetyl-2,3-dihydro, 1-acetyl-5-indolinesulfonyl chloride, n-acetylindoline-5-sulphonyl chloride, 1-acetyl-5-chlorosulfonyl indoline, 1-acetylindoline-5-sulfonylchloride, pubchem5449, acetylindolinesulfonoylchloride |
| PubChem CID | 14024596 |
| IUPAC Name | 1-acetyl-2,3-dihydroindole-5-sulfonyl chloride |
| SMILES | CC(=O)N1CCC2=C1C=CC(=C2)S(Cl)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3404403
|
Thermo Scientific Chemicals
H3404403 |
1 g |
Each for $155.70
|
|
|||||
|
AAH3404409
|
Thermo Scientific Chemicals
H3404409 |
10 g |
Each for $737.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 52206-05-0 | |
| 259.70 | |
| QNFXLCHANYHGIF-UHFFFAOYSA-N | |
| 14024596 | |
| CC(=O)N1CCC2=C1C=CC(=C2)S(Cl)(=O)=O |
| C10H10ClNO3S | |
| MFCD07368558 | |
| 1-acetyl-2,3-dihydro-1h-indole-5-sulfonyl chloride, 1-acetylindoline-5-sulfonyl chloride, 1-acetyl-5-indolinesulfonoyl chloride, 1h-indole-5-sulfonyl chloride, 1-acetyl-2,3-dihydro, 1-acetyl-5-indolinesulfonyl chloride, n-acetylindoline-5-sulphonyl chloride, 1-acetyl-5-chlorosulfonyl indoline, 1-acetylindoline-5-sulfonylchloride, pubchem5449, acetylindolinesulfonoylchloride | |
| 1-acetyl-2,3-dihydroindole-5-sulfonyl chloride |
Specifications
| 52206-05-0 | |
| C10H10ClNO3S | |
| 1 g | |
| Moisture sensitive | |
| QNFXLCHANYHGIF-UHFFFAOYSA-N | |
| 1-acetyl-2,3-dihydroindole-5-sulfonyl chloride | |
| 14024596 | |
| 97% |
| 172°C to 174°C | |
| MFCD07368558 | |
| UN3261 | |
| 1-acetyl-2,3-dihydro-1h-indole-5-sulfonyl chloride, 1-acetylindoline-5-sulfonyl chloride, 1-acetyl-5-indolinesulfonoyl chloride, 1h-indole-5-sulfonyl chloride, 1-acetyl-2,3-dihydro, 1-acetyl-5-indolinesulfonyl chloride, n-acetylindoline-5-sulphonyl chloride, 1-acetyl-5-chlorosulfonyl indoline, 1-acetylindoline-5-sulfonylchloride, pubchem5449, acetylindolinesulfonoylchloride | |
| CC(=O)N1CCC2=C1C=CC(=C2)S(Cl)(=O)=O | |
| 259.70 | |
| 259.71 | |
| 1-Acetylindoline-5-sulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P223-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only