missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylimidazole, 98%, Thermo Scientific Chemicals

Catalog No. AC169501000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169501000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC169501000 Supplier Thermo Scientific Chemicals Supplier No. 169501000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2466-76-4 | Molecular Formula: C5H6N2O | Molecular Weight: 110.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2466-76-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00005287
InChI Key VIHYIVKEECZGOU-UHFFFAOYSA-N
Synonym 1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534
PubChem CID 17174
ChEBI CHEBI:16984
IUPAC Name 1-imidazol-1-ylethanone
SMILES CC(=O)N1C=CN=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 93°C to 96°C
Color Beige to White or Tan
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ether,chloroform,thf
Formula Weight 110.12
Percent Purity 98%
Physical Form Adhering Crystals or Crystalline Powder
Chemical Name or Material 1-Acetylimidazole, 98%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
EINECSNumber 219-577-7
RTECSNumber NI3400000
TSCA TSCA

RUO – Research Use Only