missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylguanidine, 97%

CAS: 5699-40-1 | C3H7N3O | 101.11 g/mol

$182.92 - $182.92

Chemical Identifiers

CAS 5699-40-1
Molecular Formula C3H7N3O
Molecular Weight (g/mol) 101.11
MDL Number MFCD00075615
InChI Key NGGXACLSAZXJGM-UHFFFAOYSA-N
Synonym n-acetylguanidine, 1-acetylguanidine, n-carbamimidoylacetamide, n-aminoiminomethyl acetamide, monoacetyl guanidine, acetylguanidine, n-acetyl-guanidine, acetyl guanidine, n-diaminomethylidene acetamide, 1-acetylguanidne
PubChem CID 38263
IUPAC Name N-(diaminomethylidene)acetamide
SMILES CC(=O)N=C(N)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC397440050
SDS View Documents
Thermo Scientific Chemicals
397440050
5 g Glass bottle
Each for $182.92
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5699-40-1
101.11
NGGXACLSAZXJGM-UHFFFAOYSA-N
38263
CC(=O)N=C(N)N
C3H7N3O
MFCD00075615
n-acetylguanidine, 1-acetylguanidine, n-carbamimidoylacetamide, n-aminoiminomethyl acetamide, monoacetyl guanidine, acetylguanidine, n-acetyl-guanidine, acetyl guanidine, n-diaminomethylidene acetamide, 1-acetylguanidne
N-(diaminomethylidene)acetamide

Specifications

5699-40-1
100.0
White
96% min. (HPLC)
C3H7N3O
5 g
NGGXACLSAZXJGM-UHFFFAOYSA-N
N-(diaminomethylidene)acetamide
38263
97%
1-Acetylguanidine
98.5
181.0°C to 183.0°C
Authentic
Glass bottle
MFCD00075615
n-acetylguanidine, 1-acetylguanidine, n-carbamimidoylacetamide, n-aminoiminomethyl acetamide, monoacetyl guanidine, acetylguanidine, n-acetyl-guanidine, acetyl guanidine, n-diaminomethylidene acetamide, 1-acetylguanidne
CC(=O)N=C(N)N
101.11
101.11
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 227-180-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only