missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylguanidine, 97%

Catalog No. AC397440050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397440050 5 g Glass bottle
AC397440250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397440050 Supplier Thermo Scientific Chemicals Supplier No. 397440050

CAS: 5699-40-1 | C3H7N3O | 101.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5699-40-1
Molecular Formula C3H7N3O
Molecular Weight (g/mol) 101.11
MDL Number MFCD00075615
InChI Key NGGXACLSAZXJGM-UHFFFAOYSA-N
Synonym n-acetylguanidine, 1-acetylguanidine, n-carbamimidoylacetamide, n-aminoiminomethyl acetamide, monoacetyl guanidine, acetylguanidine, n-acetyl-guanidine, acetyl guanidine, n-diaminomethylidene acetamide, 1-acetylguanidne
PubChem CID 38263
IUPAC Name N-(diaminomethylidene)acetamide
SMILES CC(=O)N=C(N)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 181.0°C to 183.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 101.11
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Acetylguanidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 227-180-5

RUO – Research Use Only