missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylcyclohexene, 97%

Catalog No. p-49326
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102500050 5 g Glass bottle
AC102500250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102500050 Supplier Thermo Scientific Chemicals Supplier No. 102500050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 932-66-1 | C8H12O | 124.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 932-66-1
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.18
MDL Number MFCD00001547
InChI Key LTYLUDGDHUEBGX-UHFFFAOYSA-N
Synonym 1-acetyl-1-cyclohexene, 1-acetylcyclohexene, 1-cyclohex-1-en-1-yl ethanone, 1-cyclohexen-1-yl ethanone, ethanone, 1-1-cyclohexen-1-yl, ketone, 1-cyclohexen-1-yl methyl, unii-7539u6wqbl, methyl 1-cyclohexenyl ketone, 1-cyclohexen-1-yl methyl ketone, 1-1-cyclohexenyl-ethanone
PubChem CID 13612
IUPAC Name 1-(cyclohexen-1-yl)ethanone
SMILES CC(=O)C1=CCCCC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 73°C
Color Yellow
Density 0.9600g/mL
Boiling Point 201°C to 202°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4900 to 1.492
Linear Formula C6H9COCH3
Quantity 5 g
Beilstein 07, 58
Specific Gravity 0.96
Formula Weight 124.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Acetylcyclohexene, 97%
Show More Show Less
EINECSNumber 213-256-5

RUO – Research Use Only