missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acetyladamantane 98.0+%, TCI America™
$142.46 - $338.84
Chemical Identifiers
| CAS | 1660-04-4 |
|---|---|
| Molecular Formula | C12H18O |
| Molecular Weight (g/mol) | 178.28 |
| MDL Number | MFCD00074739 |
| InChI Key | DACIGVIOAFXPHW-UHFFFAOYSA-N |
| Synonym | 1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone |
| PubChem CID | 123126 |
| IUPAC Name | 1-(adamantan-1-yl)ethan-1-one |
| SMILES | CC(=O)C12CC3CC(CC(C3)C1)C2 |
Chemical Identifiers
| 1660-04-4 | |
| 178.28 | |
| DACIGVIOAFXPHW-UHFFFAOYSA-N | |
| 123126 | |
| CC(=O)C12CC3CC(CC(C3)C1)C2 |
| C12H18O | |
| MFCD00074739 | |
| 1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone | |
| 1-(adamantan-1-yl)ethan-1-one |
Specifications
| 1660-04-4 | |
| White-Yellow | |
| MFCD00074739 | |
| 1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone | |
| CC(=O)C12CC3CC(CC(C3)C1)C2 | |
| 178.28 | |
| 178.28 | |
| Crystalline Powder |
| 54°C | |
| C12H18O | |
| 5 g | |
| DACIGVIOAFXPHW-UHFFFAOYSA-N | |
| 1-(adamantan-1-yl)ethan-1-one | |
| 123126 | |
| ≥98.0% (GC) | |
| 1-Acetyladamantane |
Safety and Handling
TSCA : No