missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-5-bromoindole, 97%, Thermo Scientific Chemicals

Catalog No. p-7055221
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63745MD 250 mg
AAH6374503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63745MD Supplier Thermo Scientific Chemicals Supplier No. H63745MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61995-52-6
Molecular Formula C10H8BrNO
Molecular Weight (g/mol) 238.084
MDL Number MFCD00238530
InChI Key BOMKWHSZGCMFEG-UHFFFAOYSA-N
Synonym 1-acetyl-5-bromoindole, 1-acetyl-5-bromo-1h-indole, 1-5-bromo-1h-indol-1-yl ethanone, 1-5-bromoindol-1-yl ethanone, 1-5-bromo-1h-indol-1-yl ethan-1-one, ethanone, 1-5-bromo-1h-indol-1-yl
PubChem CID 12318076
IUPAC Name 1-(5-bromoindol-1-yl)ethanone
SMILES CC(=O)N1C=CC2=C1C=CC(=C2)Br

Specifications

Quantity 250 mg
Beilstein 1283925
Formula Weight 238.08
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Acetyl-5-bromoindole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only